2-Chlorobenzoylacetonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(2-Chlorophenyl)-3-oxopropanenitrile

ACTIAS34556G5EA 0 CHF
ACTIAS34556G5 ACTIAS34556G10
2-Chlorobenzoylacetonitrile
2-Chlorobenzoylacetonitrile
Formula: C₉H₆ClNO
MW: 179.61 g/mol
MDL Number: MFCD00051624
CAS Number: 40018-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR