N-Chlorophthalimide

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20922G100EA 136 CHF
ACTIAS20922G100
N-Chlorophthalimide
N-Chlorophthalimide
Formula: C₈H₄ClNO₂
MW: 181.58 g/mol
MDL Number: MFCD00023027
CAS Number: 3481-09-2
EINECS: 222-459-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR