4-Bromo-m-anisidine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS32296G5EA 0 CHF
ACTIAS32296G5 ACTIAS32296G10
4-Bromo-m-anisidine
4-Bromo-m-anisidine
Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD05664063
CAS Number: 19056-40-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR