5-Methoxy-o-toluidine
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
6-Methyl-m-anisidine
, 5-Methoxy-2-methylaniline
, 1-Amino-3-methoxy-6-methylbenzene
, 3-Amino-4-methylanisole
, 2-Amino-4-methoxytoluene
ACTIAS32391G1EA
0
CHF
ACTIAS32391G10
ACTIAS32391G1
ACTIAS32391G5
5-Methoxy-o-toluidine
5-Methoxy-o-toluidine
Formula:
C₈H₁₁NO MW: 137.18 g/mol Boiling Pt: 253 °C (1013 hPa) Melting Pt: 47 °C Density: 1.39 g/cm³ (25 °C) |
MDL Number:
MFCD00075057 CAS Number: 50868-72-9 EINECS: 256-816-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...