1-Adamantanemethanol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decaneAdamantan-1-ylmethanol1-(Hydroxymethyl)adamantane1-Adamantylmethanol

ACTIAS24522G100EA 0 CHF
ACTIAS24522G100 ACTIAS24522G25
1-Adamantanemethanol
1-Adamantanemethanol
Formula: C₁₁H₁₈O
MW: 166.26 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 115…118 °C
MDL Number: MFCD00074751
CAS Number: 770-71-8
EINECS: 212-225-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR