N-(Brommethyl)phthalimid
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
N-(Brommethyl)-1,2-benzoldicarbonsäureimid
, N-(Brommethyl)phthalsäureimid
ACTIAS23424G25EA
79
CHF
ACTIAS23424G25
N-(Brommethyl)phthalimid
N-(Brommethyl)phthalimid
Formel:
C₉H₆BrNO₂ MW: 240,06 g/mol Siedepunkt: 410 °C (1013 hPa) Melting Pt: 148…151 °C |
MDL Number:
MFCD00005897 CAS-Nummer: 5332-26-3 EINECS: 226-239-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...