2-(Trifluoromethyl)benzylamine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20417G25EA 0 CHF
ACTIAS20417G100 ACTIAS20417G25
2-(Trifluoromethyl)benzylamine
2-(Trifluoromethyl)benzylamine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 108 °C (64 mmHg)
MDL Number: MFCD00010297
CAS Number: 3048-01-9
EINECS: 221-258-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR