(Chloromethyl)trimethylsilane

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20115G100EA 0 CHF
ACTIAS20115G100 ACTIAS20115G25
(Chloromethyl)trimethylsilane
(Chloromethyl)trimethylsilane
Formula: ClH₂CSi(CH₃)₃
MW: 122.67 g/mol
Boiling Pt: 98 °C (1013 hPa)
Density: 0.8861 g/cm³ (20 °C)
MDL Number: MFCD00000878
CAS Number: 2344-80-1
EINECS: 219-058-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR