(4-Fluorphenoxy)essigsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20769G100EA 108 CHF
ACTIAS20769G100
(4-Fluorphenoxy)essigsäure
(4-Fluorphenoxy)essigsäure
Formel: (C₆H₄F)OCH₂COOH
MW: 170,14 g/mol
Melting Pt: 106 °C
Dichte: 1,516 g/cm³ (20 °C)
MDL Number: MFCD00004304
CAS-Nummer: 405-79-8
EINECS: 206-974-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR