(S)-(+)-Mandelic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: (S)-(+)-2-Hydroxy-2-phenylacetic acidL(+)-2-Hydroxy-2-phenylacetic acid(S)-2-Hydroxy-2-phenylacetic acidL(+)-Mandelic acid

ACTIAS20125G100EA 82 CHF
ACTIAS20125G100
(S)-(+)-Mandelic acid
(S)-(+)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 130…132 °C
Density: 1,349 g/cm³ (20 °C)
Flash Pt: >190 °C
MDL Number: MFCD00004495
CAS Number: 17199-29-0
EINECS: 241-240-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR