DL-Prolin

Synonyms: (±)-Prolin(±)-1,3,4,5-Tetrahydro-2H-pyrrol- 2-carbonsäure(±)-Pyrrolidin-2-carbonsäureH-DL-Pro-OH(±)-Azolidin-2-carbonsäure

ACTIAS22281G100EA 0 CHF
ACTIAS22281G100 ACTIAS22281G25
DL-Prolin
DL-Prolin
Formel: C₅H₉NO₂
MW: 115,13 g/mol
Siedepunkt: 245 °C (1013 hPa)
Melting Pt: 208…210 °C
Dichte: 1,38 g/cm³ (22.9 °C)
MDL Number: MFCD00005250
CAS-Nummer: 609-36-9
EINECS: 210-189-3
Merck Index: 12,07963

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR