(±)-3,5-Difluoromandelic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3,5-Difluoromandelic acid(±)-3,5-Difluorophenylglycolic acid

ACTIAS30562G100EA 0 CHF
ACTIAS30562G100
(±)-3,5-Difluoromandelic acid
(±)-3,5-Difluoromandelic acid
Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
MDL Number: MFCD00061300
CAS Number: 132741-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR