4-Nitropyridin-N-oxid
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS23352G100EA
0
CHF
ACTIAS23352G100
4-Nitropyridin-N-oxid
4-Nitropyridin-N-oxid
Formel:
C₅H₄N₂O₃ MW: 140,1 g/mol Siedepunkt: 397 °C (1013 hPa) Melting Pt: 161 °C Dichte: 1,52 g/cm³ (20 °C) |
MDL Number:
MFCD00006206 CAS-Nummer: 1124-33-0 EINECS: 214-395-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...