2-Acetylpyrazine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: AcetylpyrazinePyrazin-2-ylethan-1-one

ACTIAS21624G25EA 0 CHF
ACTIAS21624G25 ACTIAS21624G100
2-Acetylpyrazine
2-Acetylpyrazine
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 59 °C (33 mmHg)
Melting Pt: 79…80 °C
MDL Number: MFCD00006134
CAS Number: 22047-25-2
EINECS: 244-753-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR