1-Nitroso-2-naphthol
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
Synonyms:
2-Hydroxy-1-nitrosonaphthalin
ACTIAS24275G100EA
0
CHF
ACTIAS24275G100
1-Nitroso-2-naphthol
1-Nitroso-2-naphthol
Formel:
C₁₀H₇NO₂ MW: 173,17 g/mol Siedepunkt: 304 °C (1013 hPa) Melting Pt: 107…109 °C Lager Temperatur: Raumtemperatur |
MDL Number:
MFCD00003884 CAS-Nummer: 131-91-9 EINECS: 205-043-0 Merck Index: 12,06739 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...