1,2-Bis(diphenylphosphino)ethane

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: DIPHOSEthane-1,3-diylbis(diphenylphosphine)1,2-Bis(diphenylphosphino)ethaneEthylenebis(diphenylphosphine)Bis(1,2-diphenylphosphino)ethane

ACTIAS21625G5EA 0 CHF
ACTIAS21625G10 ACTIAS21625G5 ACTIAS21625G50
1,2-Bis(diphenylphosphino)ethane
1,2-Bis(diphenylphosphino)ethane
Formula: C₂₆H₂₄P₂
MW: 398.42 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS Number: 1663-45-2
EINECS: 216-769-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR