1-Ethylcyclohexanol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS37001G1EA 0 CHF
ACTIAS37001MG250 ACTIAS37001G1
1-Ethylcyclohexanol
1-Ethylcyclohexanol
Formula: C₈H₁₆O
Boiling Pt: 158…165 °C (1013 hPa)
Melting Pt: 34.5 °C
Density: 0.9226 g/cm³ (25 °C)
MDL Number: MFCD00021402
CAS Number: 1940-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR