ω-Bromo-4-methylacetophenone

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Bromo-4'-methylacetophenone4-Methylphenacyl bromideα-Bromo-4-methylacetophenone2-Bromo-1-(p-tolyl)ethanoneBromomethyl p-methylphenyl ketone

ACTIAS40179G100EA 0 CHF
ACTIAS40179G100
ω-Bromo-4-methylacetophenone
ω-Bromo-4-methylacetophenone
Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 105 °C (0,1 mmHg)
Melting Pt: 95…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000203
CAS Number: 619-41-0
EINECS: 210-595-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR