3-Chloro-4-iodobenzotrifluoride

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33200G5EA 0 CHF
ACTIAS33200G10 ACTIAS33200G5 ACTIAS33200G1
3-Chloro-4-iodobenzotrifluoride
3-Chloro-4-iodobenzotrifluoride
Formula: C₇H₃ClF₃I
MW: 306.45 g/mol
MDL Number: MFCD00042562
CAS Number: 141738-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR