3'-Brom-4'-fluoracetophenon
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS20864G25EA
39
CHF
ACTIAS20864G25
3'-Brom-4'-fluoracetophenon
3'-Brom-4'-fluoracetophenon
Formel:
C₈H₆BrFO MW: 217,04 g/mol Siedepunkt: 240 °C (1013 hPa) Melting Pt: 57 °C |
MDL Number:
MFCD00042466 CAS-Nummer: 1007-15-4 EINECS: 213-750-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...