4-Bromo-2-nitroaniline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20522G100EA 94 CHF
ACTIAS20522G100
4-Bromo-2-nitroaniline
4-Bromo-2-nitroaniline
Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 110…113 °C
Density: 1,81 g/cm³ (20 °C)
MDL Number: MFCD00041312
CAS Number: 875-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR