7-Aminoenanthic acid

Synonyms: 7-Aminoheptanoic acid7-Aminoheptylic acid

ACTIAS21044G5EA 0 CHF
ACTIAS21044G1 ACTIAS21044G5 ACTIAS21044G25
7-Aminoenanthic acid
7-Aminoenanthic acid
Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
MDL Number: MFCD00008242
CAS Number: 929-17-9
EINECS: 213-197-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR