Trilaurin (glycerol trilaurate)
Supplier: Thermo Scientific
Synonyms: Glyceryl tridodecanoate
J62962.14EA 132 CHF
J62962.14
Trilaurin (glycerol trilaurate)
Trilaurin (Glycerol trilaurate)
Formula: C₃₉H₇₄O₆
MW: 639.01 g/mol
Boiling Pt: 643 °C (1013 hPa)
Melting Pt: 47 °C
Density: 0.9713 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00026559
CAS Number: 538-24-9
EINECS: 208-687-0