Ytterbium trinitrate pentahydrate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Ytterbium (III) nitrate pentahydrate

ACTIAS23559G25EA 0 CHF
ACTIAS23559G25 ACTIAS23559G100
Ytterbium trinitrate pentahydrate
Ytterbium trinitrate pentahydrate
Formula: N₃O₉Yb·5H₂O
MW: 449.13 g/mol
MDL Number: MFCD00149883
CAS Number: 35725-34-9
EINECS: 237-384-6
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR