4-Bromo-2-methylthiophene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS31662G5EA 0 CHF
ACTIAS31662G5 ACTIAS31662G100 ACTIAS31662G25
4-Bromo-2-methylthiophene
4-Bromo-2-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
MDL Number: MFCD04116367
CAS Number: 29421-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR