2-Thiophenesulphonyl chloride

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: Thiophene-2-sulphonyl chloride

ACTIAS20670G25EA 0 CHF
ACTIAS20670G25 ACTIAS20670G100
2-Thiophenesulphonyl chloride
2-Thiophenesulphonyl chloride
Formula: C₄H₃ClO₂S₂
MW: 182.65 g/mol
Boiling Pt: 82…84 °C (1 mmHg)
Melting Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005426
CAS Number: 16629-19-9
EINECS: 240-677-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR