2-Bromo-5-nitrothiophene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20851G5EA 76 CHF
ACTIAS20851G25 ACTIAS20851G5
2-Bromo-5-nitrothiophene
2-Bromo-5-nitrothiophene
Formula: Câ‚„Hâ‚‚BrNOâ‚‚S
MW: 208.04 g/mol
Boiling Pt: 235…237 °C (1013 hPa)
Melting Pt: 47…48 °C
MDL Number: MFCD00022493
CAS Number: 13195-50-1
EINECS: 236-155-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR