2-Chloroisonicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Chloropyridine-4-carboxylic acid2-Chloro-4-pyridinecarboxylic acid

ACTIAS20738G100EA 0 CHF
ACTIAS20738G100
2-Chloroisonicotinic acid
2-Chloroisonicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD00191402
CAS Number: 6313-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR