6-Bromnicotinsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
6-Brompyridin-3-carbonsäure
, 6-Brom-3-pyridincarbonsäure
ACTIAS4010G5EA
0
CHF
ACTIAS4010G25
ACTIAS4010KG1
ACTIAS4010G5
6-Bromnicotinsäure
6-Bromnicotinsäure
Formel:
C₆H₄BrNO₂ MW: 202,01 g/mol |
MDL Number:
MFCD01927100 CAS-Nummer: 6311-35-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...