Chroman-4-one

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Chromanone

ACTIAS21012G25EA 0 CHF
ACTIAS21012G25 ACTIAS21012G5
Chroman-4-one
Chroman-4-one
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 39 °C
Density: 1,1291 g/cm³ (99.8 °C)
MDL Number: MFCD00006840
CAS Number: 491-37-2
EINECS: 207-736-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR