4-Phenylaniline
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Danger
|
Synonyms:
4-Aminobiphenyl
, 4-Biphenylylamine
, Biphenyl-4-ylamine
ACTIAS31139G10EA
0
CHF
ACTIAS31139G10
ACTIAS31139G5
ACTIAS31139G1
4-Phenylaniline
4-Phenylaniline
Formula:
C₁₂H₁₁N MW: 169.23 g/mol Boiling Pt: 302 °C (1013 hPa) Melting Pt: 55 °C Density: 1.16 g/cm³ (20 °C) |
MDL Number:
MFCD00007879 CAS Number: 92-67-1 EINECS: 202-177-1 Merck Index: 12,01276 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...