2-Phenylaniline

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS20438G25EA 0 CHF
ACTIAS20438G25 ACTIAS20438G100
2-Phenylaniline
2-Phenylaniline
Formula: C₁₂H₁₁N
MW: 169.23 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00007701
CAS Number: 90-41-5
EINECS: 201-990-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR