4-Ethylphenylacetylene
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS32742G5EA
0
CHF
ACTIAS32742G5
ACTIAS32742G1
ACTIAS32742G10
4-Ethylphenylacetylene
4-Ethylphenylacetylene
Formula:
C₁₀H₁₀ MW: 130.19 g/mol |
MDL Number:
MFCD00173887 CAS Number: 40307-11-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...