3,4,5-Trimethoxybenzyl alcohol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2,4,6-Trimethoxybenzyl alcohol(3,4,5-Trimethoxyphenyl)methanol

ACTIAS24585G100EA 311 CHF
ACTIAS24585G100
3,4,5-Trimethoxybenzyl alcohol
3,4,5-Trimethoxybenzyl alcohol
Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 36…38 °C
Density: 1.1427 g/cm³ (20 °C)
MDL Number: MFCD00060356
CAS Number: 3840-31-1
EINECS: 223-331-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR