4-Nitro-α,α,α-trifluoro-o-toluic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
4-Nitro-2-(trifluoromethyl)benzoic acid
ACTIAS32343G1EA
0
CHF
ACTIAS32343G1
ACTIAS32343G10
ACTIAS32343G5
4-Nitro-α,α,α-trifluoro-o-toluic acid
4-Nitro-α,α,α-trifluoro-o-toluic acid
Formula:
C₈H₄F₃NO₄ MW: 235.12 g/mol |
MDL Number:
MFCD04039246 CAS Number: 320-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...