3-Chlor-5-(trifluormethyl)benzoesäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
5-Chlor-α,α,α-trifluor-m-toluylsäure
ACTIAS35108G1EA
0
CHF
ACTIAS35108G1
ACTIAS35108G5
ACTIAS35108G10
3-Chlor-5-(trifluormethyl)benzoesäure
3-Chlor-5-(trifluormethyl)benzoesäure
Formel:
C₈H₄ClF₃O₂ MW: 224,57 g/mol |
MDL Number:
MFCD04115846 CAS-Nummer: 53985-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...