β-Chloropropiophenone

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Chloropropiophenone

ACTIAS21426G100EA 0 CHF
ACTIAS21426G100 ACTIAS21426G25
β-Chloropropiophenone
β-Chloropropiophenone
Formula: (C₆H₅)C(O)CH₂CH₂Cl
MW: 168.62 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 48…49 °C
Density: 1,181 g/cm³ (20 °C)
MDL Number: MFCD00000990
CAS Number: 936-59-4
EINECS: 213-317-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR