ω-Brom-4-fluoracetophenon
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
4-Fluorphenacylbromid
, 2-Brom-4'-fluoracetophenon
, α-Brom-4-fluoracetophenon
, 2-Brom-1-(4-fluorphenyl)ethan-1-on
, Brommethyl-p-fluorphenylketon
ACTIAS7032G5EA
0
CHF
ACTIAS7032G5
ACTIAS7032G100
ω-Brom-4-fluoracetophenon
ω-Brom-4-fluoracetophenon
Formel:
C₈H₆BrFO MW: 217,04 g/mol Siedepunkt: 82 °C (0,4 mmHg) Melting Pt: 49 °C |
MDL Number:
MFCD00040830 CAS-Nummer: 403-29-2 EINECS: 206-955-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...