4-Fluoro-3-hydroxybenzoic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS34936G5EA 0 CHF
ACTIAS34936G5 ACTIAS34936G1 ACTIAS34936G10
4-Fluoro-3-hydroxybenzoic acid
4-Fluoro-3-hydroxybenzoic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
MDL Number: MFCD00269641
CAS Number: 51446-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR