4-Fluorosalicylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40440G5EA 0 CHF
ACTIAS40440G5
4-Fluorosalicylic acid
4-Fluorosalicylic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
MDL Number: MFCD00153168
CAS Number: 345-29-9
EINECS: 206-459-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR