Methyl-3-hydroxybenzoate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Methyl 3-Hydroxybenzoate3-Hydroxybenzoic acid methyl ester

ACTIAS33009G100EA 0 CHF
ACTIAS33009G100
Methyl-3-hydroxybenzoate
Methyl 3-hydroxybenzoate
Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00002295
CAS Number: 19438-10-9
EINECS: 243-071-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR