Ethyl-3-hydroxybenzoate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-Hydroxybenzoic acid ethyl esterEthyl 3-hydroxybenzoate

ACTIAS24771G100EA 0 CHF
ACTIAS24771G100
Ethyl-3-hydroxybenzoate
Ethyl 3-hydroxybenzoate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 72…73 °C
Density: 1,068 g/cm³ (131 °C)
MDL Number: MFCD00002296
CAS Number: 7781-98-8
EINECS: 231-951-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR