Methyl-4-cyanbenzoat

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Cyanbenzoesäuremethylester

ACTIAS32799G5EA 0 CHF
ACTIAS32799G10 ACTIAS32799G5
Methyl-4-cyanbenzoat
Methyl-4-cyanbenzoat
Formel: C₉H₇NO₂
MW: 161,16 g/mol
Siedepunkt: 275 °C (1013 hPa)
Melting Pt: 67 °C
MDL Number: MFCD00001823
CAS-Nummer: 1129-35-7
EINECS: 214-443-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR