3-Bromobenzenesulphonyl chloride

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS40329G25EA 0 CHF
ACTIAS40329G25
3-Bromobenzenesulphonyl chloride
3-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
MW: 255.52 g/mol
Boiling Pt: 95…95.5 °C (0.8 mmHg)
Melting Pt: 31…33 °C
Density: 1.773 g/cm³ (20 °C)
MDL Number: MFCD00052313
CAS Number: 2905-24-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR