1-(2-Bromoethoxy)-4-nitrobenzene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35893G5EA 0 CHF
ACTIAS35893G5 ACTIAS35893G1
1-(2-Bromoethoxy)-4-nitrobenzene
1-(2-Bromoethoxy)-4-nitrobenzene
Formula: C₈H₈BrNO₃
MDL Number: MFCD00031363
CAS Number: 13288-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR