1-Bromo-4-(difluoromethoxy)benzene
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS40075G100EA
0
CHF
ACTIAS40075G100
ACTIAS40075G25
1-Bromo-4-(difluoromethoxy)benzene
1-Bromo-4-(difluoromethoxy)benzene
Formula:
C₇H₅BrF₂O MW: 223.02 g/mol |
MDL Number:
MFCD00221465 CAS Number: 5905-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...