3,3,3-Trifluoro-1-propanol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3,3,3-Trifluoropropan-1-ol

ACTIAS24897G25EA 0 CHF
ACTIAS24897G25 ACTIAS24897G5
3,3,3-Trifluoro-1-propanol
3,3,3-Trifluoro-1-propanol
Formula: C₃H₅F₃O
MW: 114.07 g/mol
Boiling Pt: 96…98 °C (1013 hPa)
Density: 1.3739 g/cm³ (25 °C)
MDL Number: MFCD00190649
CAS Number: 2240-88-2
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR