Methyl fluoroacetate

ACTIAS21697G100EA 0 CHF
ACTIAS21697G100 ACTIAS21697G25
Methyl fluoroacetate
Methyl fluoroacetate
Formula: C₃H₅FO₂
MW: 92.07 g/mol
MDL Number: MFCD00040096
CAS Number: 453-18-9
EINECS: 207-218-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR