D-Methionine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21321G100EA 0 CHF
ACTIAS21321G100 ACTIAS21321G25
D-Methionine
D-Methionine
Formula: C₅H₁₁NO₂S
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 136 °C
CAS Number: 348-67-4
EINECS: 206-483-6
Merck Index: 13,06004

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR