Methyl bromopyruvate

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Methyl 3-BromopyruvateBromopyruvic acid methyl ester3-Bromo-2-oxopropionic acid methyll ester3-Bromopyruvic acid methyll esterMethyl 3-Bromo-2-oxopropionate3-Bromo-2-oxopropionic acid methyl ester

ACTIAS51274G5EA 0 CHF
ACTIAS51274G25 ACTIAS51274G100 ACTIAS51274G5
Methyl bromopyruvate
Methyl bromopyruvate
Formula: BrH₂CC(O)COOCH₃
MW: 180.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01862963
CAS Number: 7425-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR